Assay Details
| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011785.3 | NP_035915.3 | 11 - 12 | 1297 | 149 | |
| GenBank mRNA | AF124142.1 | - | 11 - 12 | 1287 | 149 | |
| AK052953.1 | - | 3 - 4 | 409 | 149 | ||
| BC066861.1 | - | 11 - 12 | 1297 | 30089997 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akt3
Genomic Map