Assay Details
| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163433.1 | NP_001156905.1 | 1 - 2 | 300 | 89 | |
| GenBank mRNA | AK161277.1 | - | 1 - 2 | 294 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161277.1, NM_001163433.1 transcripts.
Set Membership: |
|

Samd4
Genomic Map