Assay Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080995.1 | NP_001074464.1 | 1 - 2 | 86 | 61 | |
| GenBank mRNA | AK014608.2 | - | 1 - 2 | 85 | 61 | |
| AK154915.1 | - | 1 - 2 | 68 | 61 | ||
| BC066148.1 | - | 1 - 2 | 59 | 30086532 | 61 | |
| DQ400346.1 | - | 1 - 2 | 99 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066148.1, DQ400346.1, NM_001080995.1 transcripts.
Set Membership: |
|

Ddias
Genomic Map