Assay Details
| Gene Symbol: Gpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016697.3 | NP_057906.2 | 4 - 5 | 1332 | 162 | |
| GenBank mRNA | AF185614.1 | - | 4 - 5 | 1272 | 162 | |
| AK077942.1 | - | 4 - 5 | 1310 | 162 | ||
| AK146702.1 | - | 4 - 5 | 1309 | 162 | ||
| AK159394.1 | - | 4 - 5 | 1301 | 162 | ||
| AK159435.1 | - | 4 - 5 | 1301 | 162 | ||
| AK159764.1 | - | 4 - 5 | 1309 | 162 | ||
| AK168402.1 | - | 4 - 5 | 1310 | 162 | ||
| BC036126.1 | - | 4 - 5 | 1279 | 4973409 | 162 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc3
Genomic Map