Assay Details
| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130890.2 | NP_570960.2 | 12 - 13 | 1591 | 76 | |
| GenBank mRNA | BC125504.1 | - | 12 - 13 | 1563 | 40130019 | 76 |
| BC125508.1 | - | 12 - 13 | 1563 | 40130023 | 76 | |
| BC145055.1 | - | 12 - 13 | 1563 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn8
Genomic Map