| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100014596.mm.1 |
NM_001145806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014596.mm.1 |
NM_001145806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014596.mm.1 |
NM_130890.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014596.mm.1 |
NM_130890.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014596.mm.1 |
TC0100001749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014598.mm.1 |
NM_001145806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014598.mm.1 |
NM_001145806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014598.mm.1 |
TC0100001749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014599.mm.1 |
NM_130890.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014599.mm.1 |
NM_130890.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014599.mm.1 |
TC0100001749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014597.mm.1 |
NM_001145806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014597.mm.1 |
NM_001145806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014597.mm.1 |
NM_130890.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014597.mm.1 |
NM_130890.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014597.mm.1 |
TC0100001749.mm.1 |
MTA TC |