Assay Details
| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081363.2 | NP_001074832.2 | 11 - 12 | 1704 | 130 | |
| GenBank mRNA | AK029517.1 | - | 11 - 12 | 1655 | 130 | |
| DQ642022.1 | - | 10 - 11 | 1561 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpf
Genomic Map