Assay Details
| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081363.2 | NP_001074832.2 | 12 - 13 | 5137 | 113 | |
| GenBank mRNA | AF194970.1 | - | 1 - 2 | 3422 | 113 | |
| AK157703.1 | - | 1 - 2 | 253 | 113 | ||
| DQ642022.1 | - | 11 - 12 | 4994 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpf
Genomic Map