Assay Details
| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081363.2 | NP_001074832.2 | 2 - 3 | 310 | 84 | |
| GenBank mRNA | AK029517.1 | - | 2 - 3 | 312 | 84 | |
| AK080758.1 | - | 2 - 3 | 297 | 84 | ||
| AK165236.1 | - | 2 - 3 | 311 | 84 | ||
| DQ642022.1 | - | 1 - 2 | 167 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpf
Genomic Map