Assay Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133815.2 | NP_598576.2 | 11 - 12 | 1646 | 67 | |
| GenBank mRNA | AK028426.1 | - | 11 - 12 | 1623 | 67 | |
| AK030606.1 | - | 11 - 12 | 1600 | 67 | ||
| AK033459.1 | - | 11 - 12 | 1598 | 67 | ||
| AK088179.1 | - | 11 - 12 | 1599 | 67 | ||
| AK089042.1 | - | 2 - 3 | 360 | 67 | ||
| AK151798.1 | - | 11 - 12 | 1613 | 67 | ||
| AK161762.1 | - | 11 - 12 | 1624 | 67 | ||
| AK166850.1 | - | 11 - 12 | 1586 | 67 | ||
| AK167157.1 | - | 11 - 12 | 1591 | 67 | ||
| AK203260.1 | - | 3 - 4 | 326 | 67 | ||
| BC010261.1 | - | 4 - 5 | 596 | 67 | ||
| BC014835.1 | - | 3 - 4 | 389 | 67 | ||
| BC021516.1 | - | 11 - 12 | 1587 | 5352337 | 67 | |
| BC025583.1 | - | 10 - 11 | 1395 | 67 | ||
| BC029171.1 | - | 4 - 5 | 593 | 67 | ||
| BC042522.1 | - | 11 - 12 | 1606 | 4216442 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lbr
Genomic Map