Assay Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133815.2 | NP_598576.2 | 1 - 2 | 108 | 80 | |
| GenBank mRNA | AK028426.1 | - | 1 - 2 | 85 | 80 | |
| AK030606.1 | - | 1 - 2 | 62 | 80 | ||
| AK033459.1 | - | 1 - 2 | 60 | 80 | ||
| AK088179.1 | - | 1 - 2 | 61 | 80 | ||
| AK151798.1 | - | 1 - 2 | 75 | 80 | ||
| AK161762.1 | - | 1 - 2 | 86 | 80 | ||
| AK166850.1 | - | 1 - 2 | 48 | 80 | ||
| AK167157.1 | - | 1 - 2 | 53 | 80 | ||
| BC021516.1 | - | 1 - 2 | 49 | 5352337 | 80 | |
| BC042522.1 | - | 1 - 2 | 68 | 4216442 | 80 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028426.1,AK030606.1,AK033459.1,AK151798.1,AK161762.1,AK166850.1,AK167157.1,BC021516.1,BC042522.1,NM_133815.2 transcripts.
Set Membership: |
|

Lbr
Genomic Map