Assay Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133815.2 | NP_598576.2 | 3 - 4 | 529 | 68 | |
| GenBank mRNA | AK028426.1 | - | 3 - 4 | 506 | 68 | |
| AK030606.1 | - | 3 - 4 | 483 | 68 | ||
| AK033459.1 | - | 3 - 4 | 481 | 68 | ||
| AK088179.1 | - | 3 - 4 | 482 | 68 | ||
| AK151798.1 | - | 3 - 4 | 496 | 68 | ||
| AK161762.1 | - | 3 - 4 | 507 | 68 | ||
| AK166850.1 | - | 3 - 4 | 469 | 68 | ||
| AK167157.1 | - | 3 - 4 | 474 | 68 | ||
| BC021516.1 | - | 3 - 4 | 470 | 5352337 | 68 | |
| BC025583.1 | - | 2 - 3 | 279 | 68 | ||
| BC042522.1 | - | 3 - 4 | 489 | 4216442 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lbr
Genomic Map