Assay Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145515.2 | NP_663490.2 | 6 - 7 | 571 | 104 | |
| GenBank mRNA | AB185282.1 | - | 6 - 7 | 494 | 104 | |
| AF453686.1 | - | 6 - 7 | 501 | 104 | ||
| AK173181.1 | - | 5 - 6 | 421 | 104 | ||
| BC112400.1 | - | 6 - 7 | 500 | 40059725 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mark1
Genomic Map