Assay Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145515.2 | NP_663490.2 | 8 - 9 | 866 | 71 | |
| GenBank mRNA | AB185282.1 | - | 8 - 9 | 789 | 71 | |
| AK173181.1 | - | 7 - 8 | 716 | 71 | ||
| BC112400.1 | - | 8 - 9 | 795 | 40059725 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mark1
Genomic Map