Assay Details
| Gene Symbol: Sccpdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178653.3 | NP_848768.1 | 9 - 10 | 1116 | 101 | |
| GenBank mRNA | AK042919.1 | - | 9 - 10 | 1004 | 101 | |
| AK046033.1 | - | 9 - 10 | 1117 | 101 | ||
| AK046468.1 | - | 9 - 10 | 1104 | 101 | ||
| AK050554.1 | - | 9 - 10 | 1065 | 101 | ||
| AK077695.1 | - | 9 - 10 | 1066 | 101 | ||
| AK082803.1 | - | 9 - 10 | 1001 | 101 | ||
| AK086448.1 | - | 9 - 10 | 1139 | 101 | ||
| AK166018.1 | - | 9 - 10 | 1028 | 101 | ||
| AK212321.1 | - | 3 - 4 | 266 | 101 | ||
| BC025803.1 | - | 9 - 10 | 1113 | 5100552 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sccpdh
Genomic Map