Assay Details
| Gene Symbol: Rpp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025938.4 | NP_080214.1 | 4 - 5 | 451 | 71 | |
| GenBank mRNA | AK012105.1 | - | 4 - 5 | 398 | 71 | |
| AK016631.1 | - | 4 - 5 | 398 | 71 | ||
| BC026988.1 | - | 4 - 5 | 381 | 5368565 | 71 | |
| BC046803.1 | - | 4 - 5 | 379 | 6404792 | 71 | |
| CF896465.1 | - | 4 - 5 | 451 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpp14
Genomic Map