Assay Details
| Gene Symbol: Pxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145458.3 | NP_663433.2 | 11 - 12 | 1203 | 61 | |
| NM_178279.2 | NP_840063.1 | 11 - 12 | 1203 | 61 | ||
| GenBank mRNA | AK028501.1 | - | 11 - 12 | 1153 | 61 | |
| AK048910.1 | - | 11 - 12 | 1163 | 61 | ||
| AK147037.1 | - | 11 - 12 | 1204 | 61 | ||
| AK150337.1 | - | 11 - 12 | 1188 | 61 | ||
| AK150413.1 | - | 11 - 12 | 1188 | 61 | ||
| AK150660.1 | - | 11 - 12 | 1204 | 61 | ||
| AK151098.1 | - | 11 - 12 | 1164 | 61 | ||
| AK152058.1 | - | 11 - 12 | 1162 | 61 | ||
| AK152789.1 | - | 11 - 12 | 1162 | 61 | ||
| AK153067.1 | - | 11 - 12 | 1162 | 61 | ||
| AK156146.1 | - | 11 - 12 | 1169 | 61 | ||
| AK157364.1 | - | 11 - 12 | 1197 | 61 | ||
| AK166036.1 | - | 11 - 12 | 1129 | 61 | ||
| AK169112.1 | - | 11 - 12 | 1163 | 61 | ||
| AK170361.1 | - | 11 - 12 | 1199 | 61 | ||
| AK172463.1 | - | 11 - 12 | 1204 | 61 | ||
| BC016131.1 | - | 11 - 12 | 1173 | 4922274 | 61 | |
| DQ095196.1 | - | 11 - 12 | 1101 | 61 | ||
| DQ095197.1 | - | 11 - 12 | 1101 | 61 | ||
More Information
Set Membership: |
|

Pxk
Genomic Map