Assay Details
| Gene Symbol: Pxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145458.3 | NP_663433.2 | 4 - 5 | 486 | 93 | |
| NM_178279.2 | NP_840063.1 | 4 - 5 | 486 | 93 | ||
| GenBank mRNA | AK028501.1 | - | 4 - 5 | 436 | 93 | |
| AK048910.1 | - | 4 - 5 | 446 | 93 | ||
| AK147037.1 | - | 4 - 5 | 487 | 93 | ||
| AK150337.1 | - | 4 - 5 | 471 | 93 | ||
| AK150413.1 | - | 4 - 5 | 471 | 93 | ||
| AK150660.1 | - | 4 - 5 | 487 | 93 | ||
| AK151098.1 | - | 4 - 5 | 446 | 93 | ||
| AK152058.1 | - | 4 - 5 | 445 | 93 | ||
| AK152789.1 | - | 4 - 5 | 445 | 93 | ||
| AK153067.1 | - | 4 - 5 | 445 | 93 | ||
| AK156146.1 | - | 4 - 5 | 452 | 93 | ||
| AK157364.1 | - | 4 - 5 | 480 | 93 | ||
| AK166036.1 | - | 4 - 5 | 411 | 93 | ||
| AK169112.1 | - | 4 - 5 | 446 | 93 | ||
| AK169565.1 | - | 4 - 5 | 447 | 93 | ||
| AK170361.1 | - | 4 - 5 | 482 | 93 | ||
| AK172347.1 | - | 4 - 5 | 435 | 93 | ||
| AK172463.1 | - | 4 - 5 | 487 | 93 | ||
| BC016131.1 | - | 4 - 5 | 456 | 4922274 | 93 | |
| DQ095196.1 | - | 4 - 5 | 384 | 93 | ||
| DQ095197.1 | - | 4 - 5 | 384 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pxk
Genomic Map