Assay Details
| Gene Symbol: Pxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145458.3 | NP_663433.2 | 5 - 6 | 564 | 96 | |
| NM_178279.2 | NP_840063.1 | 5 - 6 | 564 | 96 | ||
| GenBank mRNA | AK028501.1 | - | 5 - 6 | 514 | 96 | |
| AK048910.1 | - | 5 - 6 | 524 | 96 | ||
| AK147037.1 | - | 5 - 6 | 565 | 96 | ||
| AK150337.1 | - | 5 - 6 | 549 | 96 | ||
| AK150413.1 | - | 5 - 6 | 549 | 96 | ||
| AK150660.1 | - | 5 - 6 | 565 | 96 | ||
| AK151098.1 | - | 5 - 6 | 524 | 96 | ||
| AK152058.1 | - | 5 - 6 | 523 | 96 | ||
| AK152789.1 | - | 5 - 6 | 523 | 96 | ||
| AK153067.1 | - | 5 - 6 | 523 | 96 | ||
| AK156146.1 | - | 5 - 6 | 530 | 96 | ||
| AK157364.1 | - | 5 - 6 | 558 | 96 | ||
| AK166036.1 | - | 5 - 6 | 489 | 96 | ||
| AK169112.1 | - | 5 - 6 | 524 | 96 | ||
| AK169565.1 | - | 5 - 6 | 525 | 96 | ||
| AK170361.1 | - | 5 - 6 | 560 | 96 | ||
| AK172347.1 | - | 5 - 6 | 513 | 96 | ||
| BC016131.1 | - | 5 - 6 | 534 | 4922274 | 96 | |
| DQ095196.1 | - | 5 - 6 | 462 | 96 | ||
| DQ095197.1 | - | 5 - 6 | 462 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pxk
Genomic Map