Assay Details
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 16 - 17 | 2282 | 101 | |
AK052103.1 | - | 14 - 15 | 2010 | 101 | ||
AK085336.1 | - | 15 - 16 | 2157 | 101 | ||
AK086462.1 | - | 16 - 17 | 2314 | 101 | ||
BC054120.1 | - | 16 - 17 | 2267 | 6506658 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
