Assay Details
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 1 - 2 | 195 | 66 | |
AK085362.1 | - | 1 - 2 | 232 | 66 | ||
AK086462.1 | - | 1 - 2 | 225 | 66 | ||
CD348032.1 | - | 1 - 2 | 322 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
