Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 7 - 8 | 896 | 69 | |
| NR_120498.1 | 7 - 8 | 896 | 69 | |||
| GenBank mRNA | AF190325.1 | - | 7 - 8 | 712 | 69 | |
| AK042357.1 | - | 7 - 8 | 652 | 69 | ||
| AK052618.1 | - | 7 - 8 | 694 | 69 | ||
| BC069181.1 | - | 4 - 5 | 435 | 69 | ||
| BC088992.1 | - | 7 - 8 | 686 | 69 | ||
| BC099695.1 | - | 7 - 8 | 680 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lin9
Genomic Map