Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 10 - 11 | 1256 | 115 | |
| NR_120498.1 | 10 - 11 | 1254 | 115 | |||
| GenBank mRNA | AF190325.1 | - | 10 - 11 | 1072 | 115 | |
| AK052618.1 | - | 10 - 11 | 1054 | 115 | ||
| AK089989.1 | - | 10 - 11 | 1258 | 115 | ||
| BC069181.1 | - | 7 - 8 | 795 | 115 | ||
| BC088992.1 | - | 10 - 11 | 1044 | 115 | ||
| BC099695.1 | - | 10 - 11 | 1040 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lin9
Genomic Map