Assay Details
| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144796.4 | NP_659045.2 | 4 - 5 | 855 | 113 | |
| GenBank mRNA | AK035779.1 | - | 4 - 5 | 855 | 113 | |
| AK088629.1 | - | 4 - 5 | 768 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Susd4
Genomic Map