Assay Details
| Gene Symbol: Brox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027861.2 | NP_082137.1 | 4 - 5 | 418 | 107 | |
| GenBank mRNA | AK002489.1 | - | 4 - 5 | 388 | 107 | |
| AK050525.1 | - | 4 - 5 | 389 | 107 | ||
| AK162247.1 | - | 4 - 5 | 388 | 107 | ||
| BC030340.1 | - | 4 - 5 | 424 | 3498026 | 107 | |
| BC120692.1 | - | 3 - 4 | 266 | 40129615 | 107 | |
| BC125527.1 | - | 3 - 4 | 266 | 40130042 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Brox
Genomic Map