Assay Details
| Gene Symbol: Clca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178697.4 | NP_848812.1 | 9 - 10 | 1562 | 124 | |
| GenBank mRNA | AK028704.1 | - | 9 - 10 | 1563 | 124 | |
| AK035512.1 | - | 9 - 10 | 1629 | 124 | ||
| AK048276.1 | - | 9 - 10 | 1553 | 124 | ||
| AY161007.1 | - | 9 - 10 | 1518 | 124 | ||
| BC096379.1 | - | 9 - 10 | 1625 | 5317084 | 124 | |
More Information
Set Membership: |
|

Clca2
Genomic Map