Assay Details
| Gene Symbol: Elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286411.1 | NP_001273340.1 | 1 - 2 | 98 | 101 | |
| NM_001286412.1 | NP_001273341.1 | 1 - 2 | 98 | 101 | ||
| GenBank mRNA | AK132456.1 | - | 1 - 2 | 99 | 101 | |
| AK156098.1 | - | 1 - 2 | 88 | 101 | ||
| BC027532.1 | - | 1 - 2 | 66 | 101 | ||
| BC037743.1 | - | 1 - 2 | 74 | 101 | ||
| BC053914.1 | - | 1 - 2 | 60 | 101 | ||
| U19617.1 | - | 1 - 2 | 92 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132456.1, AK156098.1, BC027532.1, NM_001286411.1, NM_001286412.1, U19617.1 transcripts.
Set Membership: |
|

Elf1
Genomic Map