Assay Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014905.1 | - | 13 - 14 | 1857 | 68 | |
| AM690750.1 | - | 12 - 13 | 1498 | 68 | ||
| AM690751.1 | - | 12 - 13 | 1498 | 68 | ||
More Information
Set Membership: |
|

Ttll7
Genomic Map