Assay Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014905.1 | - | 2 - 3 | 382 | 84 | |
| CJ110550.1 | - | 2 - 3 | 421 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014905.1 transcripts.
Set Membership: |
|

Ttll7
Genomic Map