Assay Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014905.1 | - | 3 - 4 | 513 | 145 | |
| AM690750.1 | - | 2 - 3 | 154 | 145 | ||
| AM690751.1 | - | 2 - 3 | 154 | 145 | ||
More Information
Set Membership: |
|

Ttll7
Genomic Map