Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 10 - 11 | 1160 | 91 | |
| GenBank mRNA | BC041672.1 | - | 10 - 11 | 1132 | 91 | |
| BC055074.1 | - | 8 - 9 | 824 | 30052208 | 91 | |
| BC058196.1 | - | 3 - 4 | 262 | 91 | ||
| CF734341.1 | - | 5 - 6 | 539 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map