Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 11 - 12 | 1241 | 79 | |
| GenBank mRNA | BC041672.1 | - | 11 - 12 | 1213 | 79 | |
| BC055074.1 | - | 9 - 10 | 905 | 30052208 | 79 | |
| BC058196.1 | - | 4 - 5 | 343 | 79 | ||
| CF734341.1 | - | 6 - 7 | 620 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map