Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 5 - 6 | 579 | 83 | |
| GenBank mRNA | BC041672.1 | - | 5 - 6 | 551 | 83 | |
| BC055074.1 | - | 3 - 4 | 243 | 30052208 | 83 | |
| CK634659.1 | - | 5 - 6 | 465 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map