Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 9 - 10 | 1036 | 104 | |
| GenBank mRNA | BC041672.1 | - | 9 - 10 | 1008 | 104 | |
| BC055074.1 | - | 7 - 8 | 700 | 30052208 | 104 | |
| BC058196.1 | - | 2 - 3 | 138 | 104 | ||
| CF734341.1 | - | 4 - 5 | 415 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map