Assay Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033439.3 | NP_001028611.2 | 17 - 18 | 2024 | 95 | |
| NM_001252132.1 | NP_001239061.1 | 16 - 17 | 1943 | 95 | ||
| GenBank mRNA | AK076431.1 | - | 7 - 8 | 542 | 95 | |
| AK129264.1 | - | 17 - 18 | 2013 | 95 | ||
| AK136026.1 | - | 16 - 17 | 1549 | 95 | ||
| AK143909.1 | - | 16 - 17 | 1942 | 95 | ||
| AK160902.1 | - | 14 - 15 | 1319 | 95 | ||
| AK162105.1 | - | 6 - 7 | 450 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrch1
Genomic Map