Assay Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033439.3 | NP_001028611.2 | 18 - 19 | 2157 | 71 | |
| NM_001252132.1 | NP_001239061.1 | 17 - 18 | 2076 | 71 | ||
| GenBank mRNA | AK076431.1 | - | 8 - 9 | 675 | 71 | |
| AK129264.1 | - | 18 - 19 | 2146 | 71 | ||
| AK136026.1 | - | 17 - 18 | 1682 | 71 | ||
| AK143909.1 | - | 17 - 18 | 2075 | 71 | ||
| AK160902.1 | - | 15 - 16 | 1452 | 71 | ||
| AK162105.1 | - | 7 - 8 | 583 | 71 | ||
More Information
Set Membership: |
|

Lrch1
Genomic Map