Assay Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033439.3 | NP_001028611.2 | 4 - 5 | 875 | 94 | |
| NM_001252132.1 | NP_001239061.1 | 4 - 5 | 875 | 94 | ||
| GenBank mRNA | AK129264.1 | - | 4 - 5 | 864 | 94 | |
| AK136026.1 | - | 4 - 5 | 481 | 94 | ||
| AK143909.1 | - | 4 - 5 | 874 | 94 | ||
| AK160902.1 | - | 2 - 3 | 251 | 94 | ||
| AK161894.1 | - | 4 - 5 | 674 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160902.1 transcripts.
Set Membership: |
|

Lrch1
Genomic Map