Assay Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033439.3 | NP_001028611.2 | 6 - 7 | 1136 | 69 | |
| NM_001252132.1 | NP_001239061.1 | 6 - 7 | 1136 | 69 | ||
| GenBank mRNA | AK129264.1 | - | 6 - 7 | 1125 | 69 | |
| AK136026.1 | - | 6 - 7 | 742 | 69 | ||
| AK143909.1 | - | 6 - 7 | 1135 | 69 | ||
| AK160902.1 | - | 4 - 5 | 512 | 69 | ||
More Information
Set Membership: |
|

Lrch1
Genomic Map