Assay Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033439.3 | NP_001028611.2 | 7 - 8 | 1206 | 77 | |
| NM_001252132.1 | NP_001239061.1 | 7 - 8 | 1206 | 77 | ||
| GenBank mRNA | AK129264.1 | - | 7 - 8 | 1195 | 77 | |
| AK136026.1 | - | 7 - 8 | 812 | 77 | ||
| AK143909.1 | - | 7 - 8 | 1205 | 77 | ||
| AK160902.1 | - | 5 - 6 | 582 | 77 | ||
More Information
Set Membership: |
|

Lrch1
Genomic Map