Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012243.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012243.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012243.mm.1 |
TC0300001521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012244.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012244.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012244.mm.1 |
TC0300001521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012248.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012248.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012248.mm.1 |
TC0300001521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012247.mm.1 |
NM_133222.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012247.mm.1 |
NM_133222.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012247.mm.1 |
TC0300001521.mm.1 |
MTA TC |