Assay Details
| Gene Symbol: Pnldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034866.1 | NP_001030038.1 | 2 - 3 | 219 | 68 | |
| GenBank mRNA | AK139523.1 | - | 2 - 3 | 105 | 68 | |
| BC158033.1 | - | 2 - 3 | 219 | 9007336 | 68 | |
| BC172002.1 | - | 2 - 3 | 219 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139523.1 transcripts.
Set Membership: |
|

Pnldc1
Genomic Map