Assay Details
| Gene Symbol: Pnldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034866.1 | NP_001030038.1 | 5 - 6 | 452 | 103 | |
| GenBank mRNA | AK139523.1 | - | 5 - 6 | 338 | 103 | |
| BC158033.1 | - | 5 - 6 | 452 | 9007336 | 103 | |
| BC172002.1 | - | 5 - 6 | 452 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnldc1
Genomic Map