Assay Details
| Gene Symbol: Prkacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164198.1 | NP_001157670.1 | 7 - 8 | 900 | 54 | |
| NM_001164199.1 | NP_001157671.1 | 7 - 8 | 773 | 54 | ||
| NM_001164200.1 | NP_001157672.1 | 7 - 8 | 865 | 54 | ||
| NM_011100.4 | NP_035230.1 | 7 - 8 | 904 | 54 | ||
| GenBank mRNA | AK048319.1 | - | 7 - 8 | 896 | 54 | |
| AK139419.1 | - | 7 - 8 | 645 | 54 | ||
| AK148728.1 | - | 7 - 8 | 714 | 54 | ||
| AK150122.1 | - | 7 - 8 | 866 | 54 | ||
| AK150134.1 | - | 7 - 8 | 866 | 54 | ||
| AK163585.1 | - | 7 - 8 | 678 | 54 | ||
| BC054533.1 | - | 7 - 8 | 888 | 5704796 | 54 | |
| J02626.1 | - | 7 - 8 | 833 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkacb
Genomic Map