Assay Details
| Gene Symbol: Mcoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005846.2 | NP_001005846.1 | 10 - 11 | 1284 | 84 | |
| NM_026656.4 | NP_080932.2 | 10 - 11 | 1312 | 84 | ||
| GenBank mRNA | AF503575.1 | - | 10 - 11 | 1301 | 84 | |
| AK014467.1 | - | 10 - 11 | 1215 | 84 | ||
| AK019454.1 | - | 10 - 11 | 1181 | 84 | ||
| AK150446.1 | - | 10 - 11 | 1280 | 84 | ||
| AK150547.1 | - | 10 - 11 | 1280 | 84 | ||
| AK152067.1 | - | 10 - 11 | 1280 | 84 | ||
| AK152943.1 | - | 10 - 11 | 1280 | 84 | ||
| AK153290.1 | - | 10 - 11 | 1280 | 84 | ||
| AK159172.1 | - | 10 - 11 | 1312 | 84 | ||
| AY083532.1 | - | 9 - 10 | 1130 | 84 | ||
| BC029847.1 | - | 10 - 11 | 1161 | 3967456 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcoln2
Genomic Map