Assay Details
| Gene Symbol: Rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027371.3 | NP_081647.2 | 7 - 8 | 904 | 82 | |
| GenBank mRNA | AK010079.1 | - | 7 - 8 | 901 | 82 | |
| BC028802.1 | - | 1 - 2 | 139 | 82 | ||
| BC056165.1 | - | 7 - 8 | 855 | 5694049 | 82 | |
| BC089369.1 | - | 7 - 8 | 889 | 30550759 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpf1
Genomic Map