Assay Details
| Gene Symbol: Pkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178654.4 | NP_848769.2 | 9 - 10 | 1768 | 85 | |
| GenBank mRNA | AK028469.1 | - | 9 - 10 | 1755 | 85 | |
| AK046248.1 | - | 9 - 10 | 1685 | 85 | ||
| AK049713.1 | - | 9 - 10 | 1755 | 85 | ||
| AK083425.1 | - | 4 - 5 | 566 | 85 | ||
| AK133298.1 | - | 9 - 10 | 1757 | 85 | ||
| AK171092.1 | - | 9 - 10 | 1699 | 85 | ||
| BC052073.1 | - | 10 - 11 | 1691 | 6400440 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkn2
Genomic Map