Assay Details
| Gene Symbol: Arnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007488.3 | NP_031514.3 | 13 - 14 | 1548 | 61 | |
| GenBank mRNA | AB093225.1 | - | 15 - 16 | 1728 | 61 | |
| AK032628.1 | - | 13 - 14 | 1529 | 61 | ||
| BC054546.1 | - | 13 - 14 | 1469 | 6409636 | 61 | |
| D63644.1 | - | 13 - 14 | 1424 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arnt2
Genomic Map