Assay Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007402.2 | NP_031428.2 | 13 - 14 | 1365 | 66 | |
| GenBank mRNA | AF013107.1 | - | 13 - 14 | 1351 | 66 | |
| AK136692.1 | - | 13 - 14 | 1364 | 66 | ||
| KF144368.1 | - | 1 - 2 | 88 | 66 | ||
| KF144369.1 | - | 1 - 2 | 88 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam7
Genomic Map