Assay Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007402.2 | NP_031428.2 | 1 - 2 | 68 | 94 | |
| GenBank mRNA | AK136692.1 | - | 1 - 2 | 68 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_007402.2 transcripts.
Set Membership: |
|

Adam7
Genomic Map