| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400017788.mm.1 |
NM_007402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017788.mm.1 |
NM_007402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017788.mm.1 |
TC1400002301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400017789.mm.1 |
NM_007402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017789.mm.1 |
NM_007402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017789.mm.1 |
TC1400002301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400017791.mm.1 |
NM_007402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017791.mm.1 |
NM_007402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017791.mm.1 |
TC1400002301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400017787.mm.1 |
NM_007402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017787.mm.1 |
NM_007402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017787.mm.1 |
TC1400002301.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400017790.mm.1 |
NM_007402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017790.mm.1 |
NM_007402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400017790.mm.1 |
TC1400002301.mm.1 |
MTA TC |